Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Capillary columns, DB-624

on 7" cage, Capillary, DB-624, 1,80 µm, 0,32 mm, 30 m
  558-0368
 :   Agilent
Capillary columns, DB-624
  558-0368
 :  Agilent
Capillary columns, DB-624
Capillary columns, DB-624

on 7" cage, Capillary, DB-624, 1,80 µm, 0,32 mm, 30 m

 
931,00 €
   
1 * 1 ST
 

These columns are recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS volatiles analysis. They are specifically designed for the analysis of volatile priority pollutants and residual solvents.

  • Excellent inertness for active compounds
  • Bonded and cross-linked
  • Solvent rinsable
 
  • Column Format:
    Capillary
  • Film thickness:
    1,80 µm
  • Ø int.:
    0,32 mm
  • Description:
    on 7" cage
  • Length:
    30 m
  • Seq:
    47
  • Phase:
    DB-624