Order Entry
Germany
Orders LinkContactUsLinkComponent
PIPES

PIPES

Glass bottle for solids, 100 g
Catalog # BMOL52755-1
PIPES
Catalog # BMOL52755-1
CAS Number:  5625-37-6
PIPES
PIPES

Glass bottle for solids, 100 g

CAS Number:  5625-37-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
  • CAS Number:5625-37-6
  • Formula:C₈H₁₈N₂O₆S₂
  • MDL Number:MFCD00006159
  • Molecular Weight:302.37 g/mol
  • Merck Index:12,07633
  • Boiling Pt:499 °C (1013 hPa)
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:227-057-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.