MOPS-Na
Glass bottle for solids, 1 kg
Catalog # BMOL50429-2
Supplier: BIOMOL RESEARCH LABORATORIES
Jump to:
Product Details & Documents
Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
- CAS Number:71119-22-7
- Formula:C₇H₁₄NNaO₄S
- MDL Number:MFCD00064350
- Molecular Weight:231.25 g/mol
- Storage Temperature:Ambient
- EINECS:428-420-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



