Order Entry
Germany
Orders LinkContactUsLinkComponent
MOPS-Na

MOPS-Na

Glass bottle for solids, 100 g
Catalog # BMOL50429-1
MOPS-Na
Catalog # BMOL50429-1
CAS Number:  71119-22-7
MOPS-Na

Glass bottle for solids, 100 g

CAS Number:  71119-22-7
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
  • CAS Number:71119-22-7
  • Formula:C₇H₁₄NNaO₄S
  • MDL Number:MFCD00064350
  • Molecular Weight:231.25 g/mol
  • Storage Temperature:Ambient
  • EINECS:428-420-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications