Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Bis-trispropane

Glass bottle for solids, 100 g
Catalog # BMOL50224-1
Bis-trispropane
Catalog # BMOL50224-1
CAS Number:  64431-96-5
Bis-trispropane

Glass bottle for solids, 100 g

CAS Number:  64431-96-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • EINECS:264-899-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications