Orders LinkContactUsLinkComponent
D-(-)-Sorbitol
Glass bottle for solids, 500 g
  BMOL50204-1
D-(-)-Sorbitol
  BMOL50204-1
 :  50-70-4
D-(-)-Sorbitol
D-(-)-Sorbitol
Glass bottle for solids, 500 g
 :  50-70-4
1 * 500 g
 :  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  •  :50-70-4
  •  :HOH₂C(CH(OH))₄CH₂OH
  •  :MFCD00004708
  •  :182.17 g/mol
  •  :13,08797
  •  :295 °C (1013 hPa)
  •  :94…98 °C
  •  :1.524 g/cm³ (20 °C)
  •  :>100 °C
  •  :Ambient
  •  :200-061-5
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g