Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Capillary tubes

80 mm, 0,60 mm, 0,48 mm
  820-8080
 :   HILGENBERG
Capillary tubes
  820-8080
Capillary tubes
Capillary tubes

80 mm, 0,60 mm, 0,48 mm

 
54,20 €
   
1 * 100 ST
 

Soda-lime glass.

  • Both ends open
 
  • Ext.Ø:
    0,60 mm
  • Ø int.:
    0,48 mm
  • Length:
    80 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.