Microscope slide boxes
25 slides, Black/white, Wood, 25
Artikelnummer 631-9011
Hersteller: BERGMANN KARTON
Jump to:
Product Details & Documents
These boxes are designed for 25, 50 and 100 slides.
- Metal hinges and snap closure
- Numbered racks with table of contents
- Solid design with wooden frame and black coating
Specifications
- No. of slides:25
- Colour:Black/white
- Description:25 slides
- Material:Wood
- Seq:001