Measuring magnifiers
Precision scale magnifier, 23 mm
630-8271
These technical magnifiers can be used for every standard application.
- Achromatic lens made of high quality silicate for a distortion-free image with pinpoint clarity
- Fine focusing via easy-grip focusing ring
- Housing made from glass fibre reinforced polyamide
There are different precision scales available for use with the magnifiers.
: Supplied with standard crosshair scale (no. 115201), measuring range 20 mm, 1/10 mm graduation.
- Ø:23 mm
- Magnification:7×
- Description:Precision scale magnifier
- Seq:01



