Orders LinkContactUsLinkComponent
Plastic folding magnifiers
Plastic folding magnifiers
Achromatic double lens, 23 mm
Catalog # 630-8260
Supplier:   ESCHENBACH OPTIK
Plastic folding magnifiers
Catalog # 630-8260
Supplier:  ESCHENBACH OPTIK
Plastic folding magnifiers
Plastic folding magnifiers
Achromatic double lens, 23 mm
Price
148,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These magnifiers featuring a modern design and a glass fibre reinforced PA body provide a distortion-free image.
  • For handicraft and technical applications
  • Dustproof body when closed
Ø: 23 mm
Specifications
  • Ø:
    23 mm
  • Magnification:
  • Description:
    Achromatic double lens