Orders LinkContactUsLinkComponent
Reading magnifiers
Reading magnifiers
Magnifier, hand-held, 65 mm
Catalog # 630-8150
Supplier:   ESCHENBACH OPTIK
Reading magnifiers
Catalog # 630-8150
Supplier:  ESCHENBACH OPTIK
Reading magnifiers
Reading magnifiers
Magnifier, hand-held, 65 mm
Price
59,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
This magnifier provides biconvex PXM® lightweight lens for an ergonomic use and optimised performances.
  • Additional plano-convex lens in the handle for 5× magnification
  • Eyelet in the handle, round-the-neck carry cord supplied
  • Antistatic coating
Specifications
  • Ø:
    65 mm
  • Magnification:
  • Description:
    Magnifier, hand-held
  • Seq:
    02
  • Cat. No.:
    630-8150