Orders LinkContactUsLinkComponent
Pipette containers, SPECICON
Pipette containers, SPECICON
Rack, GESTICON, stainless steel, for 8 round or rectangular pipette containers (upto 70×70 mm), Dimensions 195×405×235 mm
Catalog # 612-1866
Supplier:   schuett-biotec GmbH
Pipette containers, SPECICON
Catalog # 612-1866
Pipette containers, SPECICON
Rack, GESTICON, stainless steel, for 8 round or rectangular pipette containers (upto 70×70 mm), Dimensions 195×405×235 mm
Price
146,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Containers are made of thick walled aluminium, anodised in gold colour.
  • With sterilisation opening and bayonet closures
  • Can be hermetically sealed
They can be used for the sterilisation and storage of pipettes.
Specifications
  • Description:
    Rack, GESTICON, stainless steel, for 8 round or rectangular pipette containers (upto 70×70 mm), Dimensions 195×405×235 mm
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
    Silicone insert (2 mm thick), prevents damage to pipettes caused by sudden shock, for pipette containers VARICON RS (lower part) and SPECICON (lower and upper part)
    SCUT3523213
    Log in for availability
    1 * 1 ST - 5,90 €