Orders LinkContactUsLinkComponent
Weighing scoops
Weighing scoops
60 mm, Clear
Catalog # 611-9211
Supplier:   HOFMANN GLASTECHNIK
Weighing scoops
Catalog # 611-9211
Weighing scoops
60 mm, Clear
Price
25,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Clear weighing scoops made from glass, without handle.
Specifications
  • Colour:
    Clear
  • Length:
    60 mm
  • Seq:
    01