Orders LinkContactUsLinkComponent
Scoops
160 ml, 210 mm
  231-0303
Scoops
  231-0303
Scoops
Scoops
160 ml, 210 mm
7,00 €
1 * 1 ST
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
  • Capacity:
    160 ml
  • Length:
    210 mm