Storage Bins
Blue, 315×495×185 mm
Catalog # 216-3739
Supplier: hünersdorff GmbH
Jump to:
Product Details & Documents
These storage bins are particularly suitable for a wide range of applications in industry and craft.
- Stable
- Professional quality
- Resistance to grease, resins, alkalis, oils and acids
Specifications
- Colour:Blue
- Seq:08
- W×D×H:315×495×185 mm



