Order Entry
Germany
Orders LinkContactUsLinkComponent
Storage Bins

Storage Bins

Blue, 100×115×75 mm
Catalog # 216-3735
Supplier:   hünersdorff GmbH
Storage Bins
Catalog # 216-3735
Supplier:  hünersdorff GmbH
Storage Bins
Storage Bins

Blue, 100×115×75 mm

Price
4,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

These storage bins are particularly suitable for a wide range of applications in industry and craft.

  • Stable
  • Professional quality
  • Resistance to grease, resins, alkalis, oils and acids
Specifications
  • Colour:
    Blue
  • Seq:
    01
  • W×D×H:
    100×115×75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.