Order Entry
Germany
Orders LinkContactUsLinkComponent
Measuring cylinders, conical

Measuring cylinders, conical

1500 ml, 160 mm
Catalog # 213-8022
Supplier:   CONTACTO
Measuring cylinders, conical
Catalog # 213-8022
Supplier:  CONTACTO
Measuring cylinders, conical
Measuring cylinders, conical

1500 ml, 160 mm

Price
56,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, with handle.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Capacity:
    1500 ml
  • Height:
    160 mm
  • Seq:
    03