Order Entry
Germany
Orders LinkContactUsLinkComponent
Measuring cylinders, conical

Measuring cylinders, conical

500 ml, 110 mm
Catalog # 213-8020
Supplier:   CONTACTO
Measuring cylinders, conical
Catalog # 213-8020
Supplier:  CONTACTO
Measuring cylinders, conical
Measuring cylinders, conical

500 ml, 110 mm

Price
25,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel, with handle.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Capacity:
    500 ml
  • Height:
    110 mm
  • Seq:
    01