Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Multiple socket outlets, Classic Primo

Power socket 6-way, 1,75 m (cable)
  139-1385
Multiple socket outlets, Classic Primo
  139-1385
Multiple socket outlets, Classic Primo
Multiple socket outlets, Classic Primo

Power socket 6-way, 1,75 m (cable)

 
54,82 €
   
1 * 1 ST
 

Schuko outlets in anodised aluminium, at 35° angle with nickel-plated earth and integrated child protection

  • Cable (HO5VV-F G31.50 mm²) with moulded, double-earthed right angle Schuko plug
  • Cable storage via end caps with fixing flanges
  • Two pole illuminated rocker switch

Colour: Black/silver

Dimensions: Six-way 428×72×45 mm; nine-way 556×70×45 mm

 : VDE compliant

 
  • Description:
    Power socket 6-way
  • Length:
    1,75 m (cable)
  • Seq:
    01