Order Entry
Germany
Orders LinkContactUsLinkComponent
Multiple socket outlet, six-Socket, economy

Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 5 m
Catalog # 139-1374
Supplier:   BACHMANN ELEKTRO
Multiple socket outlet, six-Socket, economy
Catalog # 139-1374
Supplier:  BACHMANN ELEKTRO
Multiple socket outlet, six-Socket, economy
Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 5 m

Price
26,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Trailing socket with built-in, attractive and practical cable rewind.

  • Moulded, shock-proof plug with double earthing
  • Mounting flange for quick, easy attachment
  • Sockets offset by 35°

Colour: White

Specifications
  • Description:
    Socket with switch and child protection, 6-way
  • Length:
    5 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.