Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 1,5 m
  139-1373
Multiple socket outlet, six-Socket, economy
  139-1373
Multiple socket outlet, six-Socket, economy
Multiple socket outlet, six-Socket, economy

Socket with switch and child protection, 6-way, 1,5 m

 
18,65 €
   
1 * 1 ST
 

Trailing socket with built-in, attractive and practical cable rewind.

  • Moulded, shock-proof plug with double earthing
  • Mounting flange for quick, easy attachment
  • Sockets offset by 35°

Colour: White

 
  • Description:
    Socket with switch and child protection, 6-way
  • Length:
    1,5 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.