Order Entry
Germany
Orders LinkContactUsLinkComponent
Extension Cables

Extension Cables

PVC extension cable, White, 5 m
Catalog # 139-1301
Supplier:   BACHMANN ELEKTRO
Extension Cables
Catalog # 139-1301
Supplier:  BACHMANN ELEKTRO
Extension Cables
Extension Cables

PVC extension cable, White, 5 m

Price
15,06 €
Log in for availability
1 * 1 ST
Product Details & Documents

PVC extension cord with moulded, double-earthed plug and Schuko coupling.

  • Cable (HO5VV-F G31,50 mm²) available in different lengths
  • Colour: white
  • Max. power: 10/16 A, 250 V~

 : VDE compliant

Specifications
  • Colour:
    White
  • Description:
    PVC extension cable
  • Length:
    5 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.