Wipes for delicate tasks, Multisoft
White, 340×320 mm, 20 packs of 25
115-2727
Soft non woven cloths made of Airlaid material, ideal for sensitive applications.
- Solvent resistant and highly absorbent
- Tear resistant, voluminous and lint-free
- Packed in convenient dispensing box for single sheet dispensing
- Packed:20 packs of 25
- Colour:White
- L×W:340×320 mm



