Orders LinkContactUsLinkComponent
Holder for wipe boxes
Holder for wipe boxes
Large holder for one wipe box, 320×127×97 mm
Catalog # 115-2106
Supplier:   HEATHROW
Holder for wipe boxes
Catalog # 115-2106
Supplier:  HEATHROW
Holder for wipe boxes
Holder for wipe boxes
Large holder for one wipe box, 320×127×97 mm
Price
60,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
These clear acrylic holders fit boxes of small or large wipes from most major brands, for example KIMWIPES*.
  • Can be wall-mounted or put on a shelf or table
  • Smaller version attaches with double-back tape, larger version needs to be mounted with screws
  • Mounting materials included
Specifications
  • Description:
    Large holder for one wipe box
  • W×D×H:
    320×127×97 mm