Orders LinkContactUsLinkComponent
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
Catalog # 531-1321
Supplier:   BUCHI
Spider evaporators
Catalog # 531-1321
Supplier:  BUCHI
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
Price
1 750,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Specifications
  • Description:
    Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
  • Type:
    For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.