Orders LinkContactUsLinkComponent
Adapters, glass, cone/screwthread
19/26 NS, GL 18, 7,5 - 9,0 mm
  201-2116
Adapters, glass, cone/screwthread
  201-2116
Adapters, glass, cone/screwthread
Adapters, glass, cone/screwthread
19/26 NS, GL 18, 7,5 - 9,0 mm
23,00 €
1 * 1 ST
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.
  • Cone size:
    19/26 NS
  • Thread:
    GL 18
  • Tube ext.Ø:
    7,5 - 9,0 mm
  • Seq:
    06