Order Entry
Germany
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

150 mm, 16 mm, 150 mm
Catalog # BRND145550
Supplier:   Brand
Funnels with short stem
Catalog # BRND145550
Supplier:  Brand
Funnels with short stem
Funnels with short stem

150 mm, 16 mm, 150 mm

Price
50,65 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

With smooth internal walls. Flare angle 60°. Funnel lip is ground ore fire-polished.

Specifications
  • Funnel O-Ø:
    150 mm
  • Stem O-Ø:
    16 mm
  • Stem length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.