Funnels with short stem
70 mm, 8 mm, 70 mm
BRND145530
: Brand
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With smooth internal walls. Flare angle 60°. Funnel lip is ground ore fire-polished.
- Funnel O-Ø:70 mm
- Stem O-Ø:8 mm
- Stem length:70 mm



