Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Diethyl ether stabilised, BAKER, J.T.Baker®

25 L
Catalog # 8106.9025
Flammable
Irritant
Supplier:   Avantor
Diethyl ether stabilised, BAKER, J.T.Baker®
Catalog # 8106.9025
Supplier:  Avantor
CAS Number:  60-29-7
Diethyl ether stabilised, BAKER, J.T.Baker®
Diethyl ether stabilised, BAKER, J.T.Baker®

25 L

CAS Number:  60-29-7
Price
528,04 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Acidity:
    passes test
  • Aldehydes:
    passes test
  • Distillation Range:
    34.0-35.0°C
  • Non-volatile Matter:
    max. 20 mg/l
  • Peroxide:
    passes test
  • Relative Density:
    0.714-0.716
  • Substances with a Foreign Odour:
    passes test
  • Water (H2O):
    max. 2 g/l

Specifications

  • Pk:
    25 L

Specifications