Diethyl ether stabilised, BAKER, J.T.Baker®
5 L
Supplier: Avantor
Diethyl ether stabilised, BAKER, J.T.Baker®
Catalog # 8106.5000
Supplier: Avantor
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Acidity:passes test
- Aldehydes:passes test
- Distillation Range:34.0-35.0°C
- Non-volatile Matter:max. 20 mg/l
- Peroxide:passes test
- Relative Density:0.714-0.716
- Substances with a Foreign Odour:passes test
- Water (H2O):max. 2 g/l
Specifications
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



