pH buffer solutions, referenced at 20 °C, Fluka™
12,00, ±0.05, Phosphate, 20 °C, Plastic bottle, 1×1 L, 1 L
Supplier: Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, Fluka™
Catalog # 33651-1L
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:1 L
- Reference temperature:20 °C
- pH accuracy:±0.05
- Pk Info:1×1 L
- pH value:12,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



