Orders LinkContactUsLinkComponent
Robotic tips for Beckman®
Robotic tips for Beckman®
918-261, 20 µl, +, -, Transparent, 10 racks, 96 each
Catalog # 736-0020
   Low Manufacturing Impact
Robotic tips for Beckman®
Catalog # 736-0020
   Low Manufacturing Impact
Robotic tips for Beckman®
Robotic tips for Beckman®
918-261, 20 µl, +, -, Transparent, 10 racks, 96 each
Price
138,00 €
Log in for availability
1 * 960 ST
Product Details & Documents
High quality tips, tested on the workstations for which they were created, to ensure compatibility and a secure fit. Certified free of RNase, DNase, DNA, RNA, pyrogens and ATP.
  • Transparent PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Low retention tips: Hydrophobic tips retain less sample which decreases the amount of reagent loss while significantly increasing result reproducibility
  • Sterile tips: Sterilised by gamma radiation
Caution: * Requires adapter 3772 only available directly from Molecular BioProducts.
Documents
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    20 µl
  • Tip style:
    918-261
  • Tip colour:
    Transparent
  • Filter:
    -
  • Cat. No.:
    736-0020
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.