Pipette controller, Pi-Pump
Pipette controller Pi-Pump, blue, for 2 ml pipettes
Catalog # 612-1925
Supplier: KARTELL
Jump to:
Product Details & Documents
Pipetting aid for all plastic and glass pipettes.
- Suction and dispensing achieved by rotating the wheel
- Liquid evacuated by pressing the lever
Specifications
- Description:Pipette controller Pi-Pump, blue, for 2 ml pipettes