Order Entry
Germany
Orders LinkContactUsLinkComponent
Hydroxylammonium sulphate ≥99%

Hydroxylammonium sulphate ≥99%

Glass bottle for solids, 500 g
Catalog # 88944.36
Health
Corrosive
Irritant
Environment
Hydroxylammonium sulphate ≥99%
Catalog # 88944.36
CAS Number:  10039-54-0
Hydroxylammonium sulphate ≥99%
Hydroxylammonium sulphate ≥99%

Glass bottle for solids, 500 g

CAS Number:  10039-54-0
Price
74,30 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 10039-54-0 Molecular Formula: H3NO.0.5H2SO4 Formula Weight: 164.14 Physical Form: Crystalline Density: 1.88 Appearance: White Odour: Odourless pH: 3.6 Melting Point: ca 170° dec. Solubility: Soluble in water Merck Reference: 14,4828 UN No.: 2865 MDL No.: MFCD00044869

Synonyms:  Bis(hydroxylammonium) sulphate, Hydroxylamine sulphate, Hydroxylamine sulphate (2:1)
  • CAS Number:10039-54-0
  • Formula:H₈N₂O₆S
  • MDL Number:MFCD00044869
  • Molecular Weight:164.14 g/mol
  • Merck Index:14,4828
  • Melting Pt:∼ 170 °C (dec.)
  • Density:1.88
  • Storage Temperature:Ambient
  • EINECS:233-118-8
  • UN:2865
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications