Orders LinkContactUsLinkComponent
Clamps, universal | Avantor
Clamps, universal
Nickel coated steel, 0 - 80 mm
Catalog # 241-7312
Supplier:   USBECK Laborgeräte
Clamps, universal
Catalog # 241-7312
Clamps, universal
Clamps, universal
Nickel coated steel, 0 - 80 mm
Wechseln zu:
Price
19,90 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents
The fingers of the clamps have cork inserts to protect the clamped items.
  • Without bosshead for fixing to stand
  • With T-screws for rapid easy clamping
Spezifikationen:
  • Spanning width:
    0 - 80 mm
  • Material:
    Nickel coated steel