Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer Solutions

9,54, ±0.01, 25 °C, 1 L
Catalog # 42423.K2
Buffer Solutions
Catalog # 42423.K2
CAS Number:  
Buffer Solutions
Buffer Solutions

9,54, ±0.01, 25 °C, 1 L

CAS Number:  
Price
44,60 €
Log in for availability
1 * 1 L
Specifications

Specifications

  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    9,54
  • Cat. No.:
    42423.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00146206

Specifications