Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer Solutions

Buffer Solutions

9,00, ±0.01, 25 °C, 500 mL
  42421.AP
Buffer Solutions
  42421.AP
 :  
Buffer Solutions
Buffer Solutions

9,00, ±0.01, 25 °C, 500 mL

 :  
 
30,70 €
   
1 * 500 mL
 

 

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    9,00
  • Cat. No.:
    42421.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00147253