Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer Solutions

Buffer Solutions

8,00, ±0.01, 25 °C, 4 L
Catalog # 42420.K7
Buffer Solutions
Catalog # 42420.K7
CAS Number:  
Buffer Solutions
Buffer Solutions

8,00, ±0.01, 25 °C, 4 L

CAS Number:  
Price
84,50 €
Log in for availability
1 * 4 L
Specifications

Specifications

  • Pk:
    4 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    8,00
  • Cat. No.:
    42420.K7
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00134130

Specifications