Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer Solutions

8,00, ±0.01, 25 °C, 1 L
  42420.K2
Buffer Solutions
  42420.K2
 :  
Buffer Solutions
Buffer Solutions

8,00, ±0.01, 25 °C, 1 L

 :  
 
47,90 €
   
1 * 1 L
 

 

  • Pk:
    1 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    8,00
  • Cat. No.:
    42420.K2
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00134130

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.