Buffer Solutions
6,00, ±0.01, 25 °C, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:6,00
- Cat. No.:42418.AP
- Physical form:Liquid
- MDL No.:MFCD00134130
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



