Buffer Solutions
5,00, ±0.01, 25 °C, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 L
- Reference temperature:25 °C
- pH accuracy:±0.01
- pH value:5,00
- Cat. No.:42417.K2
- Physical form:Liquid
- MDL No.:MFCD00147253



