Spatulas, disposable
Double spatula, with corrugated surface, 145 mm, 14 mm, White
231-1120
: SEMADENI
PS, white. These disposable spatulas are ideal for handling creams, paints or other viscous media.
- Durable, slightly elastic
- Available in different sizes and designs
- Colour:White
- Description:Double spatula, with corrugated surface
- Length:145 mm
- Seq:05
- Blade width:14 mm
- Cat. No.:231-1120
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



