Measuring spoons
95 mm, 15 ml
Catalog # 216-7352
Supplier: SEMADENI
Jump to:
Product Details & Documents
Disposable measuring spoons made of transparent PS.
Specifications
- Capacity:15 ml
- Length:95 mm
- Seq:05
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



