Bottles, serum/sample, WHEATON®
10,00 ml, 25×54 mm, Clear, No, 20 mm
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Serum bottles are ideal for long- and short-term sample storage, lyophilisation and vaccine/injectable drug containers. These moulded bottles have excellent wall strength and chemical tolerances which are important for storage applications. Bottles are manufactured from Wheaton 400 borosilicate moulded glass that conforms to USP Type I requirements. Bottles are packed in shrink-wrapped partitioned modules with the exception of 2 ml size, which has no partition.
- Fits most freeze drying applications
- Borosilicate glass conforms to USP Type I requirements
- Shrink-wrapped partitioned modules reduce breakage
Specifications
- Cap included:No
- Cap Size:20 mm
- Ø×H:25×54 mm
- Capacity:10,00 ml
- Colour:Clear
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



