Bottles, narrow neck, rectangular, PVC, with thread
500 ml, 25 mm, 70×50×189 mm
Catalog # 215-3387
Supplier: Kautex
Jump to:
Product Details & Documents
PVC, rectangular, transparent.
- Crystal clear
- Secure grip
- Leakproof
- Suitable for food
- Halal certified
Crystal clear presentation of contents, with ribbed sides for secure grip.
Specifications
- Thread:25 mm
- Capacity:500 ml
- Seq:02
- W×D×H:70×50×189 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



