Order Entry
Germany
Orders LinkContactUsLinkComponent
Measuring spoons

Measuring spoons

85 mm, 6,5 ml
Catalog # 215-0279
Supplier:   SEMADENI
Measuring spoons
Catalog # 215-0279
Supplier:  SEMADENI
Measuring spoons
Measuring spoons

85 mm, 6,5 ml

Price
31,70 €
Log in for availability
1 * 100 ST
Product Details & Documents

Disposable measuring spoons made of transparent PS.

Specifications
  • Capacity:
    6,5 ml
  • Length:
    85 mm
  • Seq:
    03