Order Entry
Germany
Orders LinkContactUsLinkComponent
Screwthread tubes, attachable

Screwthread tubes, attachable

GL 18, 16 mm, 100 mm
Catalog # 201-1602
Supplier:   witeg Labortechnik
Screwthread tubes, attachable
Catalog # 201-1602
Supplier:  witeg Labortechnik
Screwthread tubes, attachable
Screwthread tubes, attachable

GL 18, 16 mm, 100 mm

Price
2,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.

Specifications
  • Thread:
    GL 18
  • Tube ext.Ø:
    16 mm
  • Seq:
    02
  • Total length:
    100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.