Orders LinkContactUsLinkComponent
Graphite electrodes, crater
Graphite electrodes, crater
Graphite electrode, crater-drillpoint/undercut, 4,57×38,10 mm
Catalog # 40788.KN
Graphite electrodes, crater
Catalog # 40788.KN
Graphite electrodes, crater
Graphite electrode, crater-drillpoint/undercut, 4,57×38,10 mm
Price
163,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Graphite electrodes are used mainly in electric arc furnace steel production.
  • AGKSP grade
  • Ultra F purity
  • Insoluble in water
They are also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings and charging practice.
Specifications
  • Ø×L:
    4,57×38,10 mm
  • Description:
    Graphite electrode, crater-drillpoint/undercut
  • Seq:
    01