Caffeine ≥99.7%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.7% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Boiling Point: 178° subl. Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758
Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:14,1636
- Boiling Pt:178 °C (subl.)
- Melting Pt:234…236 °C
- Density:1.230
- Storage Temperature:Ambient
- EINECS:200-362-1
- UN:1544
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



