Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer Solutions

7,00, ±0.01, 25 °C, 500 mL
Catalog # 38712.AP
Buffer Solutions
Catalog # 38712.AP
CAS Number:  
Buffer Solutions
Buffer Solutions

7,00, ±0.01, 25 °C, 500 mL

CAS Number:  
Price
31,40 €
Log in for availability
1 * 500 mL
Specifications

Specifications

  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0.01
  • pH value:
    7,00
  • Cat. No.:
    38712.AP
  • Odour/scent:
    Odourless
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00146206

Specifications